siRNA Details
Gene Symbol: 2610202C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011859.1 | - | 1 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711078.1 | - | 1 | 463 | Chr. 10 - 58284431 - 58285087 [+] on Build GRCm38 |
