siRNA Details
Gene Symbol: D230034L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084380.1 | - | 1 | 3894 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB470662.2 | - | 1 | 308 | Chr. 16 - 72801390 - 72801999 [+] on Build GRCm38 |
