siRNA Details
Gene Symbol: A230001M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038386.1 | - | 1218 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB168799.2 | - | 4 | 563 | Chr. 3 - 102279093 - 102445130 [+] on Build GRCm38 |
