siRNA Details
Gene Symbol: Gm16036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139684.1 | - | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725574.1 | - | 4 | 452 | Chr. 5 - 141596425 - 141615999 [-] on Build GRCm38 |
