siRNA Details
Gene Symbol: 4930543K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019733.1 | - | 1 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715610.1 | - | 1 | 172 | Chr. 10 - 37263389 - 37263748 [-] on Build GRCm38 |
