siRNA Details
Gene Symbol: Dpysl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148950.1 | - | 1 | 382 | ||
AK149008.1 | - | 1 | 229 | |||
AK149239.1 | - | 1 | 293 |
Gene Symbol: 9430041P20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB626920.1 | - | 1 | 18 |
