siRNA Details
Gene Symbol: 1700010N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005849.1 | - | 894 | |||
AK076675.1 | - | 750 | ||||
AV260173.2 | - | 68 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA464463.1 | - | 5 | 659 | Chr. 7 - 38408163 - 38442739 [+] on Build GRCm38 |
