siRNA Details
Gene Symbol: Gm30606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039837.1 | - | 1, 2 | 218 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634395.1 | - | 1, 2 | 218 | Chr. 8 - 93707226 - 93719975 [-] on Build GRCm38 |
