siRNA Details
Gene Symbol: Gm11738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039839.1 | - | 2 | 445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634410.1 | - | 2 | 445 | Chr. 11 - 118114855 - 118116795 [+] on Build GRCm38 |
