siRNA Details
Gene Symbol: 3021401L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013907.1 | - | 1 | 743 | ||
AK034235.1 | - | 1 | 1790 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB081053.2 | - | 1 | 325 | Chr. 2 - 139673241 - 139673919 [-] on Build GRCm38 |
