siRNA Details
Gene Symbol: Gm15708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040432.1 | 3 | 1118 | |||
GenBank mRNA | AK084733.1 | - | 1 | 992 | ||
AK153727.1 | - | 3 | 1119 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040432.1 | 3 | 1118 | |||
GenBank mRNA | AK084733.1 | - | 1 | 992 | ||
AK153727.1 | - | 3 | 1119 |