siRNA Details
Gene Symbol: Gm15708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040432.1 | 3 | 400 | |||
GenBank mRNA | AK084733.1 | - | 1 | 274 | ||
AK153727.1 | - | 3 | 401 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY745687.1 | - | 3 | 398 | Chr. 5 - 144277063 - 144279373 [+] on Build GRCm38 |
