siRNA Details
Gene Symbol: Gm15246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051847.1 | - | 2164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB466597.2 | - | 1 | 146 | Chr. X - 169322440 - 169368178 [+] on Build GRCm38 |
