siRNA Details
Gene Symbol: Gm12409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046068.1 | 1 | 46 | |||
GenBank mRNA | AK041524.1 | - | 1 | 47 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB637831.1 | - | 1 | 43 | Chr. 4 - 45762375 - 45764220 [-] on Build GRCm38 |
