siRNA Details
Gene Symbol: 9530062E16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020618.1 | - | 1 | 331 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722192.1 | - | 1 | 327 | Chr. X - 130393678 - 130394321 [+] on Build GRCm38 |
