siRNA Details
Gene Symbol: Gm13261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076687.1 | - | 4 | 854 | ||
AV260557.2 | - | 4 | 487 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU605051.1 | - | 486 | Chr. - - [-] on Build |
