siRNA Details
Gene Symbol: Gm13261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076687.1 | - | 1 | 112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614999.1 | - | 1 | 109 | Chr. 2 - 10345251 - 10374039 [-] on Build GRCm38 |
