siRNA Details
Gene Symbol: Gm34753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076685.1 | - | 3 | 547 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614997.1 | - | 3 | 547 | Chr. 6 - 56369705 - 56371945 [+] on Build GRCm38 |
