siRNA Details
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039162.1 | - | 1 | 4053 | ||
BB183048.2 | - | 1 | 428 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK037387.1 | - | 437 | Chr. - - [-] on Build |
