siRNA Details
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137478.1 | - | 3 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722654.1 | - | 3 | 466 | Chr. 17 - 10840391 - 10941291 [+] on Build GRCm38 |
