siRNA Details
Gene Symbol: Pcif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009480.1 | - | 1 | 258 | ||
BC060988.1 | - | 2 | 521 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009480.1 | - | 1 | 258 | ||
BC060988.1 | - | 2 | 521 |