siRNA Details
Gene Symbol: C030015A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028820.1 | - | 1 | 2145 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB004207.2 | - | 1 | 206 | Chr. 6 - 90388736 - 90389352 [+] on Build GRCm38 |
