siRNA Details
Gene Symbol: Gm30873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035526.1 | - | 137 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB628054.1 | - | 1, 2 | 137 | Chr. 7 - 84338095 - 84347244 [+] on Build GRCm38 |
