siRNA Details
Gene Symbol: 4930578M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016308.1 | - | 2 | 386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715944.1 | - | 2 | 386 | Chr. 8 - 105859447 - 105860422 [-] on Build GRCm38 |
