siRNA Details
Gene Symbol: 2700038G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045040.1 | 4 | 650 | |||
NR_045042.1 | 4 | 631 | ||||
GenBank mRNA | AA163134.1 | - | 1 | 87 | ||
AK002390.1 | - | 4 | 651 | |||
AK012332.1 | - | 1 | 527 |
