siRNA Details
Gene Symbol: Gm15915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038017.1 | 3, 4 | 247 | |||
GenBank mRNA | AK131848.1 | - | 3, 4 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707862.1 | - | 3, 4 | 248 | Chr. 10 - 93674411 - 93683322 [-] on Build GRCm38 |
