siRNA Details
Gene Symbol: Gm36615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135696.1 | - | 3 | 640 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ052486.1 | - | 3 | 639 | Chr. 11 - 76919792 - 76930983 [-] on Build GRCm38 |
