siRNA Details
Gene Symbol: Gm40038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052977.1 | - | 2 | 507 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB603936.2 | - | 2 | 504 | Chr. 3 - 41163774 - 41172660 [-] on Build GRCm38 |
