siRNA Details
Gene Symbol: D030045P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040624.1 | 3 | 542 | |||
GenBank mRNA | AK080062.1 | - | 3 | 543 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB637151.1 | - | 3 | 543 | Chr. 10 - 45807704 - 45850736 [+] on Build GRCm38 |
