siRNA Details
Gene Symbol: Gm14025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141583.1 | - | 1, 2 | 542 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB652890.1 | - | 1, 2 | 542 | Chr. 2 - 129060946 - 129065686 [-] on Build GRCm38 |
