siRNA Details
Gene Symbol: C030011I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021065.1 | - | 1 | 431 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728141.1 | - | 1 | 431 | Chr. 17 - 90764814 - 90765485 [-] on Build GRCm38 |
