siRNA Details
Gene Symbol: 1700063H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006870.1 | - | 2 | 258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706855.1 | - | 2 | 257 | Chr. 9 - 82260294 - 82263485 [+] on Build GRCm38 |
