siRNA Details
Gene Symbol: Gm5122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040767.1 | 1 | 35 | |||
GenBank mRNA | AK040484.1 | - | 1 | 35 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB636177.1 | - | 1 | 35 | Chr. 9 - 61017069 - 61018467 [+] on Build GRCm38 |
