siRNA Details
Gene Symbol: 1700041B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018845.1 | - | 2 | 82 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706654.1 | - | 2 | 76 | Chr. 15 - 78670788 - 78676124 [+] on Build GRCm38 |
