siRNA Details
Gene Symbol: BC051226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045146.1 | 2 | 697 | |||
GenBank mRNA | AK136742.1 | - | 2 | 698 | ||
BC051226.1 | - | 2 | 424 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE629146.1 | - | 2 | 415 | Chr. 17 - 33908293 - 33908896 [-] on Build GRCm38 |
