siRNA Details
Gene Symbol: BC051226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045146.1 | 2 | 603 | |||
GenBank mRNA | AK136742.1 | - | 2 | 604 | ||
BC051226.1 | - | 2 | 330 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC051226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045146.1 | 2 | 603 | |||
GenBank mRNA | AK136742.1 | - | 2 | 604 | ||
BC051226.1 | - | 2 | 330 |