siRNA Details
Gene Symbol: 4930529L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040537.1 | 2, 3 | 210 | |||
GenBank mRNA | AK019711.1 | - | 2, 3 | 211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715499.1 | - | 2, 3 | 211 | Chr. 16 - 84679789 - 84685150 [+] on Build GRCm38 |
