siRNA Details
Gene Symbol: 1810062G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007933.1 | - | 2 | 317 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707935.1 | - | 2 | 314 | Chr. 3 - 36475940 - 36482072 [+] on Build GRCm38 |
