siRNA Details
Gene Symbol: Gm3728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053416.1 | - | 1 | 142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664962.1 | - | 1 | 142 | Chr. 19 - 20005992 - 20010191 [-] on Build GRCm38 |
