siRNA Details
Gene Symbol: Ptf1aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB701680.1 | - | 2 | 488 | ||
AK053418.1 | - | 2 | 488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB539064.2 | - | 2 | 209 | Chr. 2 - 19441508 - 19446396 [-] on Build GRCm38 |
