siRNA Details
Gene Symbol: Gm29824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052870.1 | - | 89 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661994.1 | - | 1, 2 | 89 | Chr. 9 - 24776467 - 24803378 [+] on Build GRCm38 |
