siRNA Details
Gene Symbol: Gm16348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050994.1 | - | 2 | 1547 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV046811.1 | - | 3 | 507 | Chr. 8 - 41218140 - 41220515 [+] on Build GRCm38 |
