siRNA Details
Gene Symbol: Gm34643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085825.1 | - | 3 | 2023 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB516159.2 | - | 1 | 415 | Chr. 14 - 103023777 - 103024443 [+] on Build GRCm38 |
