siRNA Details
Gene Symbol: 9530034D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020585.1 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722082.1 | - | 1 | 77 | Chr. 6 - 31330630 - 31331242 [+] on Build GRCm38 |
