siRNA Details
Gene Symbol: 1700019E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040497.1 | 4 | 381 | |||
GenBank mRNA | AK006114.1 | - | 4 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706221.1 | - | 4 | 382 | Chr. 2 - 45696605 - 45698363 [+] on Build GRCm38 |
