siRNA Details
Gene Symbol: Gm30173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040671.1 | - | 1 | 99 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB636576.1 | - | 1 | 99 | Chr. 3 - 52516964 - 52549780 [+] on Build GRCm38 |
