siRNA Details
Gene Symbol: Gm30173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040671.1 | - | 5 | 622 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA718882.1 | - | 3 | 387 | Chr. 3 - 52543472 - 52549793 [+] on Build GRCm38 |
