siRNA Details
Gene Symbol: 1700003K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005648.1 | - | 1 | 359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705839.1 | - | 1 | 359 | Chr. 14 - 71946794 - 71947213 [+] on Build GRCm38 |
