siRNA Details
Gene Symbol: Gm568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA867168.1 | - | 2 | 429 | ||
BC028561.1 | - | 2 | 429 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX525764.1 | - | 2 | 427 | Chr. 4 - 47008316 - 47010779 [-] on Build GRCm38 |
