siRNA Details
Gene Symbol: Gm38758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062895.1 | - | 1 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU704146.1 | - | 1 | 49 | Chr. 5 - 150603816 - 150606994 [-] on Build GRCm38 |
